3,3'-Diaminobenzidin

Lieferant: MP Biomedicals
Gefahr

Synonyme: DAB, Biphenyl-3,3',4,4'-tetrayltetraamin

08980681
ICNA08980681EA 305 EUR
ICNA08980681
3,3'-Diaminobenzidin
3,3'-Diaminobenzidin

Storage temperature: -20°C


pH 4.94 (1 tab/10mL H2O)

Formel: C₁₂H₁₄N₄
Molecular Weight: 214,27 g/mol
MDL: MFCD00007725
CAS-Nummer: 91-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR