Cell culture media, powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Lieferant: MP Biomedicals
091003122
ICNA091003122EA 45.2 EUR
ICNA091003122
Cell culture media, powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Zellkulturmedien
Basal Medium Eagle is suited for the isolation and cultivation of non-fastidious and fastidious microorganisms from a variety of clinical and non-clinical specimens. It can also be used as a selective isolation medium by adding antimicrobial agents.