Dehydrated culture media, potato dextrose agar, MP Biomedicals
Lieferant: MP Biomedicals
Potato dextrose agar (PDA) is suited for the cultivation of yeasts and moulds from dairy products and other foods. It may also be used to induce sporulation in many fungi.
Standards: This medium is recommended by the Association of Public Health (APHA).