Dehydrated culture media, tryptic soy broth, MP Biomedicals
Lieferant: MP Biomedicals
Tryptic Soy Broth is used for the cultivation of fastidious and non-fastidious microorganisms. It is a highly nutritive and versatile medium in the laboratory. This medium promotes abundant growth of fastidious microorganisms without the necessity of adding serum and other materials. It may also be used for the rapid determination of the bacteriological quality of water.
- Tan powder format
- pH 7,1 to 7,5
- Stored at room temperature
- Ideal for bacteria cell culture
Storage temperature : Room temperature (15...30 °C)
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...