Dehydrated culture media, terrific broth, MP Biomedicals
Lieferant: MP Biomedicals
091012017
ICNA091012017EA
87.8
EUR
ICNA091012017
Dehydrated culture media, terrific broth, MP Biomedicals
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
This dehydrated medium is used for molecular biology applications.
- Ideal for bacteria cell culture
- To be stored at room temperature