Kanamycinsulfat Lösung
Lieferant: MP Biomedicals
Synonyme:
Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
091672048
ICNA091672048EA
68.8
EUR
ICNA091672048
Kanamycinsulfat Lösung
Kanamycin sulphate
Kanamycin is an aminoglycoside antibiotic effective against mycoplasma, gram-positive and gram-negative bacteria. It is used in biotechnology applications to inhibit protein synthesis.
- Binds to 70S ribosomal subunit
- Inhibits translocation
- Elicits miscoding
Formel:
C₁₈H₃₈N₄O₁₅S Molecular Weight: 582,6 g/mol |
MDL:
MFCD00070253 CAS-Nummer: 25389-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...