L(+)-Glutamin
Lieferant: MP Biomedicals
Synonyme:
L(+)-Glutaminsäure-5-amid, Q, (S)-(+)-2-Amino-1,5-pentandisäure-5-amid, (S)-(+)-Glutamin, L(+)-2-Amino-1,5-pentandisäure-5-amid, H-Gln-OH, (S)-(+)-Glutaminsäure-5-amid, Gln, Levoglutamid
091680149
ICNA091680149EA
26.7
EUR
ICNA091680149
L(+)-Glutamin
L(+)-Glutamin
Formel:
C₅H₁₀N₂O₃ Molecular Weight: 146.15 g/mol Siedepunkt: 355 |
MDL:
MFCD00008044 CAS-Nummer: 56-85-9 Merck Index: 13,04484 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...