Lymphocyte separation medium, LymphoSep™, MP Biomedicals
Lieferant: MP Biomedicals
091692249 091692254
ICNA091692249EA 53.1 EUR
ICNA091692249 ICNA091692254
Lymphocyte separation medium, LymphoSep™, MP Biomedicals
Zelltrennmedien
Lymphocyte separation medium, LYMPHOSEP® is a sterile, iso-osmotic, low viscosity solution for density centrifugation yielding a clear separation of lymphocytes from heparinised or defibrinated human blood.
  • In vitro isolation of lymphocytes from diluted whole blood
  • Maximum yield of mononuclear cells
  • Easy one-step centrifugation
  • >96% cell viability of lymphocytes
  • Low endotoxins
  • Autoclave sterilization
Lymphocytes are isolated by layering diluted, heparinized blood over a solution of sodium metrizoate that contains a polysaccharide. This polysaccharide aggregates the erythrocytes so that they can be sedimented by low centrifugation. The less dense lymphocytes remain as a clearly defined band at the plasma LymphoSep interface.
Bitte beachten Store at room temperature (15 to 30 °C), protect from light.