TCM® Defined Serum Replacement, MP Biomedicals
Lieferant: MP Biomedicals
092010026
ICNA092010026EA 218 EUR
ICNA092010026
TCM® Defined Serum Replacement, MP Biomedicals
Zellkulturmedien
This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
  • A universal serum replacement that supports human and mammalian cells
  • Completely free of virus, mycoplasma and other infectious agents
  • Contains no growth factors, steroid hormones, extraneous proteins or phenol red
  • Protein origin - human; Drug Master on file with the FDA
  • Recommended for use in the production of cell-secreted proteins
TCM™ is a fortified, multipurpose serum replacement for long-term culturing of many types of anchorage dependent and suspension cultures. Highly effective in the production of biologicals, as well as in culturing other mammalian cells.