TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedicals
Lieferant: MP Biomedicals
This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
- A universal serum replacement that supports human and mammalian cells
- Completely free of virus, mycoplasma and other infectious agents
- Contains no growth factors, steroid hormones, extraneous proteins or phenol red
- Protein origin - human; Drug Master on file with the FDA
TCH is a fortified, multipurpose serum replacement for long-term culturing of primarily human anchorage-dependent and suspension cells. Highly effective in the production of biologicals, as well as in culturing bacteria cell culture and other mammalian cells.