EDTA disodium salt solution, 0,2% in phosphate buffered saline, MP Biomedicals
Lieferant: MP Biomedicals
092820549
ICNA092820549EA
24
EUR
ICNA092820549
EDTA disodium salt solution, 0,2% in phosphate buffered saline, MP Biomedicals
Puffer
Klonpuffer
EDTA disodium salt, (2%) solution, in phosphate buffered saline without calcium and magnesium. Used as a gentle non-enzymatic cell dissociation reagent.
- Clear and colourless
- pH: 4,0 to 6,0
- Osmolality: 240 to 450 mM/kg