Fetal Bovine Serum, Gold, Heat Inactivated, MP Biomedicals
Lieferant: MP Biomedicals
092916849
ICNA092916849EA 0 EUR
ICNA092916849
Fetal Bovine Serum, Gold, Heat Inactivated, MP Biomedicals
Seren
Fetal bovine serum has been the most widely used growth supplement for cell culture media because of its high content of embryonic growth-promoting factors.
  • Supplies components that satisfy specific metabolic requirements for the culture of cells in vitro
  • Suitable for cell culture and insect cell culture
  • Source of growth promoting and survival enhancing factor
CELLect™ FBS, Gold, Heat Inactivated is used in a broad range of cell culture applications.
Serum is an extremely complex mixture of many small and large biomolecules with different, physiologically balanced growth-promoting and growth-inhibiting activities. Some of the major functions of serum are to provide growth factors, hormones, attachment and spreading factors, binding proteins, lipids, and minerals.
FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.