Multitest slide, MP Biomedicals
Lieferant: MP Biomedicals
096041805E
ICNA096041805EEA 202 EUR
ICNA096041805E
Multitest slide, MP Biomedicals
Objektträger Kammerobjektträger
These high quality glass slides have a unique PTFE coating, which offers several important advantages.
  • Self draining surface
  • Resistant to solvents and fixatives
  • Resistant to direct heat, sterilisable
Storage temperature: 15 to 30 °C