(2-Nitrophenyl)-β-D-galactopyranosid (ONPG), kristallin

Lieferant: MP Biomedicals

Synonyme: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

02102473.5 10247301 10247305 0210247301 0210247305 0210247325
ICNA02102473.5EA 31.7 EUR
ICNA02102473.5 ICNA10247301 ICNA10247305 ICNA0210247301 ICNA0210247305 ICNA0210247325
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG), kristallin
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG)

o-Nitrophenyl-β-D-galactoside is a cell culture component for molecular genetics. It is chromogenic substrate for b-galactosidase and hence is useful in the detection of lacZ activity.


β-galactosidase, breaks down lactose into galactose and glucose. β-Galactosidase is not lactose specific and can act on simple galactosides. 2-Nitrophenyl β-D-galactopyranoside hydrolysis results in the release of galactose and a yellow chromogenic compound. The test substrate does not depend on an induced or constitutive permease enzyme to enter the cell, therefore reactions are rapid and occur within a 24-hour period.


β-galactosidase, breaks down lactose into galactose and glucose. β-Galactosidase is not lactose specific and can act on simple galactosides. 2-Nitrophenyl β-D-galactopyranoside hydrolysis results in the release of galactose and a yellow chromogenic compound. The test substrate does not depend on an induced or constitutive permease enzyme to enter the cell, therefore reactions are rapid and occur within a 24-hour period.


Soluble in water (10 mg/ml- clear, colorless solution; 15 mg/ml - clear, faint yellow solution; 20 mg/ml with heat - clear, yellowish solution).


Bitte beachten Research Use Only (RUO).

Formel: C₁₂H₁₅NO₈
Lagertemperatur: Kühlschrank
MDL: MFCD00063255
CAS-Nummer: 369-07-3

Bestellen

Spezifikation Testergebnisse

Identity test Passes
FTIR Conforms to standard
Optical Rotation (c=1, H₂O) –66° ± 7°
Melting Point 180 - 195 °C
UV/VIS: λmax (H₂O) 262 ±3 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR