RNA Purification kits, RNAID® spin kit, MP Biomedicals
Lieferant: MP Biomedicals
The RNaid® Spin Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels. The kit includes spin filters which prevent silica particle carry-over into cleaned RNA.
Ideal for removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions. The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.