Dehydrated culture media, SOC medium, MP Biomedicals
Lieferant: MP Biomedicals
113031012
ICNA113031012EA 86.3 EUR
ICNA113031012
Dehydrated culture media, SOC medium, MP Biomedicals
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
SOC medium is a rich media used primarily in the recovery step of Escherichia coli competent cell transformations.
  • Maximises the transformation efficiency of competent cells
SOC powder is a special medium for incubating competent cells immediately after transformation to allow expression of transferred resistance genes before exposing cells to selective conditions.
Bestellinformation: Small quantities of biological buffers have been added for proper pH adjustment.