Dehydrated culture media, Terrific broth I & II, MP Biomedicals
Lieferant: MP Biomedicals
113046041 113045032 113046022 113046032 113046042 113046052
ICNA113046041EA 1583 EUR
ICNA113046041 ICNA113045032 ICNA113046022 ICNA113046032 ICNA113046042 ICNA113046052
Dehydrated culture media, Terrific broth I & II, MP Biomedicals
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Terrific broth (TB) has been formulated for optimum growth of Escherichia coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using this broth.
  • Molecular biology certified bacterial medium
Terrific broth I is shipped as two separate components, part one contains 24 g yeast extract, 12 g tryptone & part 2 contains potassium phosphate salts and glycerol in sterile solution.
Terrific Broth II is a proprietary formulation that is shipped as a single powder instead of as two separate components exhibiting same growth characteristics as the original terrific broth formula.