Fast RNA™ spin kit for microbes, MP Biomedicals
Lieferant: MP Biomedicals
116020050
ICNA116020050EA 651 EUR
ICNA116020050
Fast RNA™ spin kit for microbes, MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The FastRNA™ SPIN Kit for Microbes quickly and efficiently isolates high-quality, total RNA from tough-to-lyse bacterial cell cultures and other microbial sources in approximately 15 minutes using a zirconium silicate Lysing Matrix (0,1 mm) to lyse bacteria and SPIN columns for the purification process.
The FastRNA™ SPIN Kit isolates both large and small RNA species without the use of phenol or reducing agents. Small RNAs (eg. tRNAs, microRNAs) can be recovered following a simple adjustment within the RNA isolation protocol. RNA (~5 µg) isolated from 10¹ to 10⁵ cells can be eluted into volumes as little as 6 µl in less than 15 minutes. High quality total RNA (A260/A280 >1,8, A260/A230 >1,8) is recovered. RNA is eluted with RNase-free water and can be stored at –70 °C. The addition of RNase inhibitors is optional, but highly recommended for prolonged storage.
For research purposes only.
Lieferumfang: Kit contains Lysing Matrix B (0,1 mm zirconium silicate), 50×2 ml tubes; FastRNA™ Lysis Buffer, 50 ml; FastRNA™ Prep Buffer, 25 ml; FastRNA™ Wash Buffer, 12 ml; Water (DNase/RNase-free), 1 ml; FastRNA™ Spin Column 1, 50 each; FastRNA™ Spin Column 2, 50 each; and Collection Tubes (2 ml), 100 each.