Fast RNA™ Pro Soil-Direct Kit, MP Biomedicals
Lieferant: MP Biomedicals
116070050
ICNA116070050EA 589 EUR
ICNA116070050
Fast RNA™ Pro Soil-Direct Kit, MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
FastRNA™ Pro Soil-Direct Kit is used to isolate total RNA from organic material found in soil samples. The total RNA isolated is suitable for RT-PCR analysis and other downstream applications.
For research purposes only.
Lieferumfang: Kit contains RNApro™ Soil Lysis Solution, 55 ml; Phenol:Chloroform (1:1), 50 ml; Inhibitor Removal Solution, 12 ml; RNAMATRIX® Slurry, 0,6 ml; RNAMATRIX® Binding Solution, 35 ml; RNAMATRIX® Wash Solution Concentrate, 15 ml; DEPC-treated H2O, 30 ml; Lysing Matrix E, 50×2 ml tubes; and Quick-Clean Spin Filters.