Fast DNA™ SPIN kit, MP Biomedicals
Lieferant: MP Biomedicals
The Fast DNA® SPIN Kit is used with the FastPrep®-24 or FastPrep® FP120 instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.
- Isolation of genomic DNA from plants, animals, bacteria, yeast, algae, and fungi using a silica spin filter method
- Rapid and reproducible sample lysis with the FastPrep®-24 or FastPrep® FP120 instrument
- Lyse and isolate DNA in less than 30 minutes from a variety of sample types
- Flexible format eliminates need for separate DNA isolation kits
- No hazardous organic reagents required
- SPIN filters are included to streamline silica handling
Lysing Matrix A and three different chaotropic buffers are provided for the optimal homogenisation of a wide variety of sample types (plant and animal tissues, cultured cells, bacteria, yeast, fungi, insects, etc). The released DNA is purified by a silica-based GeneClean® SPIN procedure, with supplied filter buckets and catch tubes to simplify liquid handling, and is suitable for PCR analysis and other downstream applications.
For research purposes only.
Lieferumfang: Kit contains Lysing Matrix A, 100×2,0 ml tubes;1/4" Ceramic Spheres, 100 spheres; Binding Matrix, 66 ml; Concentrated SEWS-M, 12 ml; DES, 25 ml; CLS-VF, 90 ml; PPS, 25 ml; CLS-TC, 110 ml; CLS-Y, 110 ml; SPIN Modules, 100 each; and Catch Tubes, 100 each.