Nucleic acid purification system, MPure-12™, MP Biomedicals

Lieferant: MP Biomedicals
117002200
ICNA117002200EA 24309 EUR
ICNA117002200
Nucleic acid purification system, MPure-12™, MP Biomedicals
Automatisierte Aufreinigungssysteme für Nukleinsäure
MPure-12™ is a bench top automated system for rapid purification of nucleic acids from a wide variety of bio-specimens using magnetic bead separation technology. It has a uniquely designed magnetic bead processing chamber and is fully integrated with easy to use pre-packaged reagent kits.

  • Fully automated and integrated platform that offers true walk-away operation
  • Purify nucleic acids of the highest yield and purity, ready for immediate use in downstream applications
  • Flexible in throughput, process 1 to 12 samples from a wide variety of biological specimens in a single run
  • Uniquely designed Polygon Reaction Chamber minimises cross contamination and mag bead carryover
  • Simple operation, Bar-code Scanner quickly enters program parameters in seconds
  • Minimize nucleic acid loss and degradation
  • Reproducible, sample-to-sample consistency, scalable and convenient
  • Closed system that offers increased safety and efficiency

MPure-12™ is a state of the art bench-top automated nucleic acid purification system that rapidly provides superior yield and purity of nucleic acids for any downstream application including qPCR, RE digestion and Southerns. The MPure-12 system utilizes a unique magnetic bead separation technology, is fully integrated with easy to use pre-packaged reagent kits and provides superior nucleic acid purification results from a wide variety of biological specimens.

Nucleic acids extracted can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.

Specifications
Sample Type: Plasma, Serum, Blood, Plants, Cultured Cells, Bacteria, FFPE, Forensic samples, Swabs, Tissues
Capacity: Purify Nucleic acids from 100 to 1000 µl of sample
Sensitivity: As little as 5 IU spiked (about 1 IU in PCR reaction) sample can be detected, proving the excellent sensitivity and linearity of isolation
Time setting: 30 to 70 minutes
Dimensions: 48W×70D×52H cm
Weight: 43 kg
Power requirements: 110 to 240 V, 50/60 Hz

Bitte beachten Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR