DNA extraction kit, forensic, MPure™ 12, MP Biomedicals
Lieferant: MP Biomedicals
117022110
ICNA117022110EA
335
EUR
ICNA117022110
DNA extraction kit, forensic, MPure™ 12, MP Biomedicals
Nukleinsäure-Reagenzien
Reinigungskits und Reagenzien für Nukleinsäure
MPure™ Forensic DNA Extraction Kit is used with the MPure-12™ aNAP system for the extraction and purification of genomic DNA from forensic samples.