Nucleic acid extraction, MPure™ Viral/Pathogen nucleic acids extraction kit, MP Biomedicals
Lieferant: MP Biomedicals
117022130
ICNA117022130EA 335 EUR
ICNA117022130
Nucleic acid extraction, MPure™ Viral/Pathogen nucleic acids extraction kit, MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
MPure™ Viral/Pathogen Nucleic Acids Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for purification of viral DNA/RNA and pathogen DNA from swab samples (cell-rich samples).