DNA extraction kits, MPure™ Plant DNA extraction kit, MP Biomedicals
Lieferant: MP Biomedicals
117022150
ICNA117022150EA 335 EUR
ICNA117022150
DNA extraction kits, MPure™ Plant DNA extraction kit, MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
MPure™ Plant DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction of genomic DNA from plant (leaf, seeds and spores) and fungal tissues. Up to 100 mg of tissue can be used for purification.