RNA extraction, MPure™ Total RNA extraction kit , MP Biomedicals
Lieferant: MP Biomedicals
117022160
ICNA117022160EA 338 EUR
ICNA117022160
RNA extraction, MPure™ Total RNA extraction kit , MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
MPure™ Total RNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction of total RNA from whole blood, blood cells, animal tissue, plant tissue, yeast or cultured cells and viral RNA from human biological specimens such as serum, plasma, and other cell-free fluids.