DNA extraction kits, MPure™ Bacterial DNA extraction kit, MP Biomedicals
Lieferant: MP Biomedicals
117022600
ICNA117022600EA
335
EUR
ICNA117022600
DNA extraction kits, MPure™ Bacterial DNA extraction kit, MP Biomedicals
Nukleinsäure-Reagenzien
Reinigungskits und Reagenzien für Nukleinsäure
MPure™ Bacterial DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction and purification of genomic DNA from both gram-positive and gram-negative bacteria. Nucleic acids extracted and purified from MPure™ Bacterial DNA Extraction Kit can be used in a number of downstream application including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis.