DNA extraction, MPure™ FFPE DNA extraction kit, MP Biomedicals
Lieferant: MP Biomedicals
117022900
ICNA117022900EA
335
EUR
ICNA117022900
DNA extraction, MPure™ FFPE DNA extraction kit, MP Biomedicals
Nukleinsäure-Reagenzien
Reinigungskits und Reagenzien für Nukleinsäure
MPure™ FFPE DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for the extraction and purification of genomic DNA from FFPE (formalin-fixed, paraffin-embedded) tissue samples, providing high quality, high integrity DNA for molecular diagnostics and research use.