Agarose, QA-Agarose™, niedrigschmelzend

Lieferant: MP Biomedicals

11AGAL0050
ICNA11AGAL0050EA 230 EUR
ICNA11AGAL0050
Agarose, QA-Agarose™, niedrigschmelzend
Agarose

Agarose low melting point is an intermediate melting and gelling point agarose which exhibits superior sieving characteristics. It is recommended for analytical gels of DNA fragments under 1000 bp and is ideal for small DNA fragments down to 20 bp can be separated on 6 % gels. It resolves fragments differing in size by only 2 bp. Its use avoids the need for PAGE Southern and Northern transfers can be achieved efficiently from them at concentrations up to 2%. DNA fragments can be purified with high efficiency from this agarose gels up to 4% by using either a purification kit or an electroelution system.

Agarose Low Melting Point is an intermediate melting agarose with approximately twice the resolution capability of routine agarose. It is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae. It is ideal for the resolution of small DNA fragments in the range 20 bp to 2000 bp.

Trade Name
QA-Agarose™

Lagertemperatur: Raumtemperatur
MDL: MFCD00081294
CAS-Nummer: 9012-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR