Rubber cement, fixogum, MP Biomedicals
Lieferant: MP Biomedicals
11FIXO0125
ICNA11FIXO0125EA 80.6 EUR
ICNA11FIXO0125
Rubber cement, fixogum, MP Biomedicals
Klebstoffe
Fixogum Rubber Cement is made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridisation for FISH.