Vacuum blotter, TransDNA Express™, MP Biomedicals
Lieferant: MP Biomedicals
11INSB8100
ICNA11INSB8100EA 4104 EUR
ICNA11INSB8100
Vacuum blotter, TransDNA Express™, MP Biomedicals
Blotting-Apparate
TransDNA Express™ vacuum blotter is used to quickly and efficiently transfer DNA and RNA to membranes. Transfer is completed within an hour without any additional handling.
  • Easy to use
  • Reproducible
  • Transfers with no gel collapse