dNTP set, MP Biomedicals
Lieferant: MP Biomedicals
11NTACG111
11NTACG100
ICNA11NTACG111EA
532
EUR
ICNA11NTACG111
ICNA11NTACG100
dNTP set, MP Biomedicals
Nukleinsäure-Reagenzien
Nukleotide
Set consists of 100 mM aqueous solutions of dATP, dCTP, dGTP and dTTP each supplied in a separate vial.