Carbenicillin Dinatriumsalz, frei rinnendes Pulver
Lieferant: MP Biomedicals
|
Gefahr
|
Synonyme:
NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
Semi-synthetic antibiotic related to penicillin. It is a benzyl penicillin derivative with substitution of a-carboxyl group n the benzyl side chain. The product interferes with final cell wall synthesis of susceptible bacteria. It is completely ionized in the blood. Carbencillin disodium is about 50% reversibly protein bound in plasma and its tissue distribution is similar to other penicillins.
- Presentation: Free Flowing Powder
- pKa: (in water) 3.3
- Soluble in water (50 mg/ ml) and alcohol
hough Carbencillin disodium has substantial in vitro activity against a variety of both gram-positive and gram-negative microorganisms, the most important aspect of its profile is in its antipseudomonal and antiproteal effect.
Used for selection of ampr transformed cells and to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.
Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Analog to ampicillin. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.
Formel:
C₁₇H₁₆N₂O₆S·2Na Lagertemperatur: Kühlschrank |
MDL:
MFCD00077683 CAS-Nummer: 4800-94-6 EINECS: 225-360-8 |
Spezifikation Testergebnisse
Identity Test | Passes |
Potency | >650 µg/mg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...