Rotationsverdampfer System RV 3 pro V-C EUComplete 1 * 1 ST
Lieferant: IKA JANKE KUNKEL
Evaporator, Rotary, Vertical cooling, Temperature range: Ambient… 99 °C, Rotation speed: 20 - 300 min⁻¹, W×D×H: 440×530×330 mm, Weight: 28,3 kg