Homogenisers, T 25 digital LR
Lieferant: IKA
20008820
IKAA20008820EA 2669 EUR
IKAA20008820
Homogenisers, T 25 digital LR
Homogenisatoren Tischhomogenisatoren
Powerful disperser for volumes ranging from 1 to 2000 ml (H2O) with digital speed display. The wide speed range of 600 to 25000 rpm facilitates work with high circumferential speeds. An extensive selection of dispersing shafts makes multiple applications possible. The application range includes homogenisation and dispersion tasks for use under vacuum in the IKA laboratory reactors.
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Quick and easy cleaning of stainless steel dispersing shafts
  • Error code display
  • Quiet operation
Standards: IP protection class according to DIN EN 60529: IP 20
Bestellinformation: Dispersing elements and accessories need to be ordered separately.