Vortex mixer, VORTEX 2
Lieferant: IKA
25000258
IKAA25000258EA 302 EUR
IKAA25000258
Vortex mixer, VORTEX 2
Schüttler Vortex-Schüttler
Vortex shaker with sturdy metal die cast housing. Suitable for continuous operation (low heat due to ventilation of motor).
  • Stable operation at high speeds due to silicon feet
  • Infinitely adjustable speed range
  • Temperature range 5 to 40 °C
  • Various applications are possible because of 3 interchangeable attachments and 7 inserts
  • Touch function
Standards: IP protection class according to DIN EN 60529: IP 21
Bestellinformation: Supplied without accessories. A range of attachments is available, these must be ordered separately.