Sentinel® Back Harness
Lieferant: ILC DOVER
S-3105
ILCDS-3105EA 0 EUR
ILCDS-3105
Sentinel® Back Harness
Atemschutzmasken Atemschutz-Systeme
The ergonomic back harness permits the Sentinel® blower to be worn comfortably on the back.
  • Ergonomic design
  • Blower securely snaps into the back harness
  • Supports longer shift operations
The harness features heat-sealed construction for easy cleaning, and is ideal for use throughout long shifts.