Sentinel XT® Clear Full Hood
Lieferant: ILC DOVER
S-3112-30
ILCDS-3112-30EA 0 EUR
ILCDS-3112-30
Sentinel XT® Clear Full Hood
Respirators Atemschutz-Systeme
The Sentinel XT® Clear Full Hood is a superior solution for protection in the pharmaceutical industry when compared to other PAPR masks and facepieces.
  • For use with the Sentinel XT® Blower system
  • 320 degree field of view
  • No fit test required
  • Save time and storage space with just one size needed
  • APF of 1000
  • Latex-free
With an APF of 1000, the clear full hood offers a 320 degree field of view, allowing the wearer to have facial hair and/or a head covering, and reducing feelings of claustrophobia. No headband or ratchet harness is needed to secure the hood, ensuring a more comfortable experience. Metal clips allow the outer bib to secure to protective clothing.