Sentinel® Extendible Breathing Tube
Lieferant: ILC DOVER
S-3506
ILCDS-3506EA 0 EUR
ILCDS-3506
Sentinel® Extendible Breathing Tube
Atemschutzmasken Atemschutz-Systeme
With minimum length of 48 cm, this flexible breathing tube can be extended to 72 cm to accommodate a wide range of heights and movements.
  • Flexible breathing tube
  • Easily extends to support a wide range of motions