Sentinel® Quick-Loc Belt
Lieferant: ILC DOVER
S-4011
ILCDS-4011EA 0 EUR
ILCDS-4011
Sentinel® Quick-Loc Belt
Atemschutzmasken Atemschutz-Systeme
Easy to clean waist belt quickly and securely snaps into the ergonomically designed Sentinel® blowers for hands-free operation.
  • Blower quickly snaps into the belt
  • Easily cleaned with wipes or immersion
Belt is designed to contour to the user's back for increased comfort.