Sentinel XE® LiPO Battery Charging Kit
Lieferant: ILC DOVER
S-4504EU
ILCDS-4504EUEA 0 EUR
ILCDS-4504EU
Sentinel XE® LiPO Battery Charging Kit
Atemschutzmasken Atemschutz-Systeme
Charges Sentinel XE® LiPO rechargeable battery.
  • Easy to use charger
  • Battery slides in and securely engages
  • Available with EU or UK plug adapter
Charging takes approximately 5 hours if battery is completely drained. Contains cradle and plug adapter.