Welch® Laboratory Vacuum Systems
Lieferant: Welch by Gardner Denver
116264
ILMV116264EA 0 EUR
ILMV116264
Welch® Laboratory Vacuum Systems
Pumpen Rotationsverdampfer-Pumpen
Chemically resistant diaphragm pump with frequency changer and gas ballast valve. Suction-side separator with 500 ml round-bottomed flask.
  • Pressure-side, insulated intensive cooler with round-bottom flask 500 ml with safety valve
  • Vacuum control box VCB 521 ef with sensor and venting valve
  • One controller regulated connection
  • Suction connection DN 16 (DN 8 optional)
  • Assembled and wired ready for connection