tempmate.®-GS2 Single-Use Temperature Data Logger
Lieferant: tempmate
G2H-000
IMECG2H-000EA 131 EUR
IMECG2H-000
tempmate.®-GS2 Single-Use Temperature Data Logger
Datenlogger
The tempmate.®-GS2 offers you the possibility to perform the worldwide* real-time monitoring of all relevant data. By setting individual alarm limits, you are alerted in real-time. Damage can therefore be clearly localised and identified. With continuous use, you can optimise your transport routes and internal processes.
  • Flexible, carefree and simple handling
  • Automatically collect data in real-time
  • See the measurements on a free cloud interface
  • Carry out in-depth analyses of your data
  • Dimensions: 103×64×23 mm for Lithium and 103×64×30 mm for Non-Lithium
  • Weight: 130 g for Lithium and 175 g for Non-Lithium
The tempmate.®-GS2 offers a wide range of configuration options to flexibly design each device according to your requirements. You can rely on being alerted by an alarm in real-time and track your consignment very closely via the tempmate.®-cloud.
With your tempmate.®-GS2, you are safe in the knowledge that your goods are in safe hands round-the-clock. The tempmate.®-GS2 not only monitors the temperature of your goods, but is also your reliable partner in humidity, light incidence, localisation and extreme temperature ranges from −200 to +100 °C. And data loss is no longer a problem either thanks to the USB export feature. So you can hand over highly sensitive goods without any worries.
One touch of a button and your consignment is in good hands. After starting the tempmate.®-GS2, the device automatically starts sending regular updates about the condition of your goods to the free tempmate.®-cloud. Even registration and configuration of individual devices are done in a few simple steps and you can quickly concentrate your valuable time on your daily tasks again without security falling by the wayside.
Standards:
EN12830, EMC, RoHS, FCC, IP 65.