Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: ImmunoReagents
GTXTK-003DHRPX
IMMRGTXTK-003DHRPXEA 128 EUR
IMMRGTXTK-003DHRPX
Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Turkey