Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: ImmunoReagents
IR2222
IMMRIR2222EA 91.6 EUR
IMMRIR2222
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Standards: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Bestellinformation: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR