Anti-IgG Goat Polyclonal Antibody (DyLight® 633)

Lieferant: ImmunoReagents
IR2323
IMMRIR2323EA 294 EUR
IMMRIR2323
Anti-IgG Goat Polyclonal Antibody (DyLight® 633)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR