Orbital shakers, benchtop, orbitron
Lieferant: INFORS
63967
INFO63967EA 4669 EUR
INFO63967
Orbital shakers, benchtop, orbitron
Schüttler Kreisschüttler
Orbitron is a shaker for demanding continuous operation, even with maximum load. This extraordinarily stable, splash-proof shaker is suitable both for daily operations in the lab and for use in climate-controlled rooms.
  • Corrosion resistant and watertight
  • Rounded edges make it easy to clean
  • Quiet and vibration-free
  • Dynamic mass-balancing system
  • Precise shaking speed control
  • Splash-proof display
  • Displays shaking speed and time with integrated timer
This benchtop shaker can be loaded with up to 31 kg of materials. The clever quick-lock system changes trays in no time.
Standards: In accordance with IP43 - protected against objects larger than 1 mm and against spraying water.