Super Safe sampler
Lieferant: INFORS
65373
INFO65373EA 0 EUR
INFO65373
Super Safe sampler
Probenehmer Probenehmer für Flüssigkeiten
This sampler allows you to take smallest samples without wasting any culture.
  • Sterile sampling without laminar flow hood
  • Truly aseptic and prevents faulty operation and contamination
  • More precise results especially in small scale bioprocesses – especially important for small culture vessels and parallel bioprocess
Can be used for all types of small culture vessels like bags, spinner flasks, stirred tank reactors.