Filter Bags, BagPage Plus®
Lieferant: INTERSCIENCE
122025 123010 121025 161025 122425 122325
432-0008EA 297 EUR
432-0008 INSC123010 INSC121025 INSC161025 INSC122425 INSC122325
Filter Bags, BagPage Plus®
Beutel Homogenisationsbeutel
BagPage® single use bags with filter membrane reduce analysis time thanks to their wide opening and markings showing where to introduce the pipette. Ideal for pasty products, cheese, flour and cakes. To use: Place the sample in the opened compartment (Fig. 1). Mix and then add reagents into the specially designed compartment (Fig. 2).
  • Strong with wide opening for introducing the sample and easy pipetting
  • Pipettes are not blocked by food fragments, which are retained behind the filter membrane
  • Transparent and rigid with a full page micro perforated,
More Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable