Storage rack for bags, BagRack® Slide
Lieferant: INTERSCIENCE
221440
INSC221440EA 226 EUR
INSC221440
Storage rack for bags, BagRack® Slide
Gestelle Beutelständer
BagRack® Slide enables the clean and methodical storage of 12 bags with samples. The BagRack® Slide allows the user to slide the bag from the side for insertion and also for access for pipetting.
  • All stainless steel
  • Autoclavable
  • Stackable
This rack is compatible with pre-enrichment of the sample for the detection of pathogens bacteria such as listeria or salmonella.
Compatible with:
- BagFilter® Pipet & Roll for easy pipetting and quick and clean disposal, avoids cross contaminations from the pipet
- BagFilter® Roll for quick and clean disposal
- BagClip® and BagFilter® Pipet for easy pipetting
- BagClip® and standard 400 blender bags