Peristaltic pumps, FlexiPump®

Lieferant: INTERSCIENCE
561000 562000
INSC561000EA 3860 EUR
INSC561000 INSC562000
Peristaltic pumps, FlexiPump®
Pumpen Peristaltikpumpen
FlexiPump® is a range of peristaltic pumps for culture media preparation. Use it for serial dispensing of culture media, agar or any liquid diluent for efficient sample preparation.

  • Select from 3 dispensing modes: Continuous, dose (predetermined volume), multi-dose (predetermined volume, with an adjustable pause between each dose)
  • 32 editable programs
  • Rotating button for easy navigation
  • Compact: Only 16 cm wide
  • Precise flow control: Up to 1% precision
  • Dispenses liquid from 50 μl to 99 L
  • Contamination free: Dispensing assembly is entirely autoclavable with
More Product Information
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR